Assay Details
Gene Symbol: NAT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266409.1 | NP_001253338.1 | 259 | 73 | ||
XM_015120241.2 | XP_014975727.1 | 346 | 73 | |||
XM_015120242.2 | XP_014975728.1 | 343 | 73 | |||
XM_015120243.2 | XP_014975729.1 | 498 | 73 | |||
XM_015120244.2 | XP_014975730.1 | 321 | 73 | |||
XM_015120245.2 | XP_014975731.1 | 365 | 73 | |||
XM_028835494.1 | XP_028691327.1 | 343 | 73 | |||
XM_028835495.1 | XP_028691328.1 | 350 | 73 | |||
XM_028835496.1 | XP_028691329.1 | 327 | 73 | |||
GenBank mRNA | JU333928.1 | - | 194 | 73 | ||
JV046722.1 | - | 259 | 73 | |||
JV711775.1 | - | 134 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
