Assay Details
| Gene Symbol: YTHDF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265926.1 | NP_001252855.1 | 2037 | 82 | ||
| GenBank mRNA | JU334895.1 | - | 1874 | 82 | ||
| JU475901.1 | - | 1916 | 82 | |||
| JV043553.1 | - | 2037 | 82 | |||
| JV470922.1 | - | 1761 | 82 | |||
| JV653353.1 | - | 2038 | 82 | |||
More Information
Set Membership: |
|

YTHDF2
Genomic Map