Assay Details
| Gene Symbol: SCPEP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261026.1 | NP_001247955.1 | 1005 | 84 | ||
| XM_028835554.1 | XP_028691387.1 | 1109 | 84 | |||
| GenBank mRNA | JU326629.1 | - | 970 | 84 | ||
| JU476652.1 | - | 1015 | 84 | |||
| JV047831.1 | - | 1005 | 84 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SCPEP1
Genomic Map