Assay Details
| Gene Symbol: PRKCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143833.2 | XP_014999319.1 | 2235 | 57 | ||
| GenBank mRNA | JU327440.1 | - | 595 | 57 | ||
| JU471201.1 | - | 1881 | 57 | |||
More Information
Set Membership: |
|

PRKCH
Genomic Map