Assay Details
| Gene Symbol: WEE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114885.2 | XP_014970371.2 | 1098 | 92 | ||
| XM_028833694.1 | XP_028689527.1 | 1129 | 92 | |||
| XM_028833695.1 | XP_028689528.1 | 1129 | 92 | |||
| GenBank mRNA | JU492863.1 | - | 428 | 92 | ||
More Information
Set Membership: |
|

WEE1
Genomic Map