Assay Details
| Gene Symbol: BLM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097543.4 | XP_001097543.2 | 2994 | 82 | ||
| XM_015143029.2 | XP_014998515.2 | 2994 | 82 | |||
| XM_015143030.2 | XP_014998516.2 | 1803 | 82 | |||
| XM_028852012.1 | XP_028707845.1 | 3049 | 82 | |||
| GenBank mRNA | JU476758.1 | - | 2913 | 82 | ||
More Information
Set Membership: |
|

BLM
Genomic Map