Assay Details
| Gene Symbol: MALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086151.4 | XP_001086151.1 | 2004 | 80 | ||
| XM_002800951.3 | XP_002800997.1 | 1971 | 80 | |||
| GenBank mRNA | JU479160.1 | - | 1605 | 80 | ||
| JU479162.1 | - | 1572 | 80 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MALT1
Genomic Map