Assay Details
| Gene Symbol: CD38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261773.1 | NP_001248702.1 | 784 | 67 | ||
| XM_028848132.1 | XP_028703965.1 | 903 | 67 | |||
| GenBank mRNA | JU337205.1 | - | 822 | 67 | ||
| JU477923.1 | - | 784 | 67 | |||
| JV727416.1 | - | 846 | 67 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CD38
Genomic Map