Assay Details
| Gene Symbol: MAPRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266800.1 | NP_001253729.1 | 238 | 82 | ||
| XM_028827333.1 | XP_028683166.1 | 143 | 82 | |||
| GenBank mRNA | JU321913.1 | - | 138 | 82 | ||
| JU470977.1 | - | 245 | 82 | |||
| JV045112.1 | - | 238 | 82 | |||
| JV637506.1 | - | 261 | 82 | |||
More Information
Set Membership: |
|

MAPRE1
Genomic Map