Assay Details
| Gene Symbol: RPL10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266226.1 | NP_001253155.1 | 42 | 59 | ||
| GenBank mRNA | JV528297.1 | - | 42 | 59 | ||
| JV600169.1 | - | 42 | 59 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266226.1 transcripts.
Set Membership: |
|

RPL10A
Genomic Map