Assay Details
| Gene Symbol: TAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148644.2 | XP_015004130.2 | 3012 | 70 | ||
| GenBank mRNA | JU492280.1 | - | 1980 | 70 | ||
| JV555983.1 | - | 108 | 70 | |||
More Information
Set Membership: |
|

TAF4
Genomic Map