Assay Details
| Gene Symbol: PDE6H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151202.2 | XP_015006688.1 | 654 | 78 | ||
| XM_015151203.2 | XP_015006689.1 | 1207 | 78 | |||
| GenBank mRNA | JU487944.1 | - | 113 | 78 | ||
More Information
Set Membership: |
|

PDE6H
Genomic Map