Assay Details
| Gene Symbol: HERPUD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095112.3 | XP_001095112.2 | 699 | 72 | ||
| XM_015126223.1 | XP_014981709.1 | 699 | 72 | |||
| GenBank mRNA | JU471638.1 | - | 238 | 72 | ||
| JV045651.1 | - | 360 | 72 | |||
| JV602557.1 | - | 487 | 72 | |||
| JV638999.1 | - | 385 | 72 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HERPUD1
Genomic Map