Assay Details
| Gene Symbol: RBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194541.1 | NP_001181470.1 | 521 | 114 | ||
| XM_028828999.1 | XP_028684832.1 | 788 | 114 | |||
| GenBank mRNA | JU470957.1 | - | 362 | 114 | ||
More Information
Set Membership: |
|

RBP5
Genomic Map