Assay Details
| Gene Symbol: HARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141041.2 | XP_014996527.1 | 652 | 84 | ||
| GenBank mRNA | JU334238.1 | - | 473 | 84 | ||
| JU475356.1 | - | 488 | 84 | |||
| JV046897.1 | - | 455 | 84 | |||
More Information
Set Membership: |
|

HARS1
Genomic Map