Assay Details
| Gene Symbol: GPAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145309.2 | XP_015000795.2 | 1609 | 86 | ||
| XM_015145310.2 | XP_015000796.2 | 1497 | 86 | |||
| XM_028852621.1 | XP_028708454.1 | 552 | 86 | |||
| GenBank mRNA | JU320614.1 | - | 1520 | 86 | ||
| JU471026.1 | - | 1547 | 86 | |||
| JV045609.1 | - | 1538 | 86 | |||
| JV637464.1 | - | 1549 | 86 | |||
More Information
Set Membership: |
|

GPAT4
Genomic Map