Assay Details
| Gene Symbol: SCD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193798.1 | NP_001180727.1 | 881 | 68 | ||
| GenBank mRNA | JU322588.1 | - | 856 | 68 | ||
| JV043242.1 | - | 864 | 68 | |||
| JV567600.1 | - | 885 | 68 | |||
| JV602574.1 | - | 883 | 68 | |||
| JV638267.1 | - | 875 | 68 | |||
| JV669035.1 | - | 880 | 68 | |||
More Information
Set Membership: |
|

SCD5
Genomic Map