Assay Details
| Gene Symbol: TENT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082136.4 | XP_001082136.2 | 1460 | 109 | ||
| XM_001082391.4 | XP_001082391.1 | 920 | 109 | |||
More Information
Set Membership: |
|

TENT5A
Genomic Map