Assay Details
| Gene Symbol: GNPTG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204748.2 | NP_001191677.2 | 432 | 94 | ||
| XM_015125251.2 | XP_014980737.2 | 565 | 94 | |||
| GenBank mRNA | EG704989.1 | - | 384 | 94 | ||
| JU325470.1 | - | 344 | 94 | |||
| JV565194.1 | - | 420 | 94 | |||
| JV658925.1 | - | 2376 | 94 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GNPTG
Genomic Map