Assay Details
| Gene Symbol: RCBTB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260614.1 | NP_001247543.1 | 1060 | 58 | ||
| XM_015120904.2 | XP_014976390.1 | 1139 | 58 | |||
| XM_015120905.2 | XP_014976391.1 | 1033 | 58 | |||
| GenBank mRNA | JU476231.1 | - | 1060 | 58 | ||
| JU476232.1 | - | 1060 | 58 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RCBTB1
Genomic Map