Assay Details
| Gene Symbol: CPEB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097847.4 | XP_001097847.3 | 3328 | 67 | ||
| XM_001097943.4 | XP_001097943.2 | 3352 | 67 | |||
| XM_001098122.4 | XP_001098122.2 | 3344 | 67 | |||
| XM_015141429.2 | XP_014996915.1 | 2045 | 67 | |||
| GenBank mRNA | JU319891.1 | - | 3270 | 67 | ||
| JU336501.1 | - | 3246 | 67 | |||
| JU477312.1 | - | 2137 | 67 | |||
| JV724094.1 | - | 3246 | 67 | |||
More Information
Set Membership: |
|

CPEB4
Genomic Map