Assay Details
| Gene Symbol: ADAMTSL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028838643.1 | XP_028694476.1 | 1168 | 58 | ||
| XM_028838644.1 | XP_028694477.1 | 1181 | 58 | |||
| XM_028838645.1 | XP_028694478.1 | 814 | 58 | |||
| XM_028838646.1 | XP_028694479.1 | 1168 | 58 | |||
| GenBank mRNA | JV713679.1 | - | 814 | 58 | ||
More Information
Set Membership: |
|

ADAMTSL5
Genomic Map