Assay Details
Gene Symbol: WDR38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193696.3 | NP_001180625.2 | 845 | 59 | ||
XM_015116852.2 | XP_014972338.2 | 625 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
