Assay Details
| Gene Symbol: NRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147113.2 | XP_015002599.2 | 1297 | 69 | ||
| XM_015147114.2 | XP_015002600.2 | 1297 | 69 | |||
| XM_015147115.2 | XP_015002601.2 | 1372 | 69 | |||
| XM_015147116.2 | XP_015002602.2 | 1297 | 69 | |||
| XM_015147117.2 | XP_015002603.2 | 1297 | 69 | |||
| XM_015147119.2 | XP_015002605.2 | 1297 | 69 | |||
| XM_015147120.2 | XP_015002606.2 | 1294 | 69 | |||
| XM_028826401.1 | XP_028682234.1 | 1375 | 69 | |||
| XM_028826402.1 | XP_028682235.1 | 1375 | 69 | |||
| XM_028826403.1 | XP_028682236.1 | 1375 | 69 | |||
| XM_028826404.1 | XP_028682237.1 | 1375 | 69 | |||
| XM_028826406.1 | XP_028682239.1 | 1375 | 69 | |||
| XM_028826407.1 | XP_028682240.1 | 1294 | 69 | |||
| GenBank mRNA | JU327093.1 | - | 807 | 69 | ||
More Information
Set Membership: |
|

NRG3
Genomic Map