Assay Details
| Gene Symbol: ARID5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093968.4 | XP_001093968.2 | 1833 | 140 | ||
| XM_015147540.2 | XP_015003026.2 | 776 | 140 | |||
| XM_028826602.1 | XP_028682435.1 | 954 | 140 | |||
| XM_028826603.1 | XP_028682436.1 | 1602 | 140 | |||
| XM_028826604.1 | XP_028682437.1 | 1102 | 140 | |||
| GenBank mRNA | JU336669.1 | - | 1121 | 140 | ||
| JU477469.1 | - | 1117 | 140 | |||
| JV724917.1 | - | 987 | 140 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ARID5B
Genomic Map