Assay Details
| Gene Symbol: ACP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123889.2 | XP_014979375.1 | 1008 | 90 | ||
| XM_015123890.2 | XP_014979376.1 | 958 | 90 | |||
| XM_015123891.2 | XP_014979377.1 | 994 | 90 | |||
| GenBank mRNA | JU321238.1 | - | 914 | 90 | ||
| JU321239.1 | - | 914 | 90 | |||
More Information
Set Membership: |
|

ACP7
Genomic Map