Assay Details
| Gene Symbol: KCNN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110811.2 | XP_014966297.1 | 91 | |||
| XM_015110839.2 | XP_014966325.1 | 91 | ||||
| GenBank mRNA | JV714449.1 | - | 774 | 91 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

KCNN3
Genomic Map