Assay Details
| Gene Symbol: KIF3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266930.1 | NP_001253859.1 | 2181 | 60 | ||
| GenBank mRNA | JU321598.1 | - | 2181 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIF3C| Gene Symbol: KIF3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266930.1 | NP_001253859.1 | 2181 | 60 | ||
| GenBank mRNA | JU321598.1 | - | 2181 | 60 | ||
Set Membership: |
|