Assay Details
| Gene Symbol: CLPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839865.1 | XP_028695698.1 | 300 | 68 | ||
| GenBank mRNA | JU322856.1 | - | 234 | 68 | ||
| JU471569.1 | - | 212 | 68 | |||
| JV669144.1 | - | 230 | 68 | |||
More Information
Set Membership: |
|

CLPP
Genomic Map