Assay Details
| Gene Symbol: NDUFS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194303.2 | NP_001181232.1 | 587 | 63 | ||
| XM_028832695.1 | XP_028688528.1 | 661 | 63 | |||
| GenBank mRNA | JU334648.1 | - | 569 | 63 | ||
| JU471124.1 | - | 542 | 63 | |||
| JV061888.1 | - | 458 | 63 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NDUFS3
Genomic Map