Assay Details
| Gene Symbol: NPRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099730.4 | XP_001099730.2 | 1504 | 63 | ||
| XM_015130842.2 | XP_014986328.1 | 1520 | 63 | |||
| XM_015130843.2 | XP_014986329.1 | 1502 | 63 | |||
| XM_015130845.2 | XP_014986331.1 | 1430 | 63 | |||
| XM_015130846.2 | XP_014986332.1 | 1718 | 63 | |||
| XM_015130850.2 | XP_014986336.1 | 1191 | 63 | |||
| XM_015130851.2 | XP_014986337.1 | 1147 | 63 | |||
| XM_028844170.1 | XP_028700003.1 | 1400 | 63 | |||
| GenBank mRNA | JU321861.1 | - | 1224 | 63 | ||
| JU472777.1 | - | 1221 | 63 | |||
| JV044413.1 | - | 1203 | 63 | |||
| JV667651.1 | - | 1217 | 63 | |||
More Information
Set Membership: |
|

NPRL2
Genomic Map