Assay Details
| Gene Symbol: LDB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266417.1 | NP_001253346.1 | 653 | 61 | ||
| GenBank mRNA | JU336929.1 | - | 449 | 61 | ||
| JU336930.1 | - | 449 | 61 | |||
| JU477666.1 | - | 622 | 61 | |||
| JU477667.1 | - | 622 | 61 | |||
| JV048594.1 | - | 653 | 61 | |||
| JV048595.1 | - | 653 | 61 | |||
| JV700127.1 | - | 669 | 61 | |||
More Information
Set Membership: |
|

LDB1
Genomic Map