Assay Details
| Gene Symbol: LTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047150.1 | NP_001040615.1 | 210 | 143 | ||
| GenBank mRNA | JU486647.1 | - | 77 | 143 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LTB
Genomic Map