Assay Details
| Gene Symbol: PAX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126048.2 | XP_014981534.2 | 536 | 55 | ||
| XM_015126203.2 | XP_014981689.1 | 533 | 55 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAX7| Gene Symbol: PAX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126048.2 | XP_014981534.2 | 536 | 55 | ||
| XM_015126203.2 | XP_014981689.1 | 533 | 55 | |||
Set Membership: |
|