Assay Details
| Gene Symbol: NPAS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143520.2 | XP_014999006.1 | 719 | 78 | ||
| XM_015143521.2 | XP_014999007.1 | 2364 | 78 | |||
| XM_015143523.2 | XP_014999009.1 | 634 | 78 | |||
| XM_015143524.2 | XP_014999010.1 | 532 | 78 | |||
| XM_028851475.1 | XP_028707308.1 | 2415 | 78 | |||
| XM_028851477.1 | XP_028707310.1 | 2415 | 78 | |||
| XM_028851478.1 | XP_028707311.1 | 686 | 78 | |||
| XM_028851479.1 | XP_028707312.1 | 2358 | 78 | |||
| XM_028851480.1 | XP_028707313.1 | 2363 | 78 | |||
| XM_028851481.1 | XP_028707314.1 | 628 | 78 | |||
| XM_028851482.1 | XP_028707315.1 | 622 | 78 | |||
| XM_028851483.1 | XP_028707316.1 | 577 | 78 | |||
| XM_028851484.1 | XP_028707317.1 | 571 | 78 | |||
| XM_028851486.1 | XP_028707319.1 | 577 | 78 | |||
| GenBank mRNA | JV478230.1 | - | 515 | 78 | ||
More Information
Set Membership: |
|

NPAS3
Genomic Map