Assay Details
| Gene Symbol: TTC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266162.2 | NP_001253091.1 | 513 | 97 | ||
| GenBank mRNA | JU322911.1 | - | 513 | 97 | ||
| JV518901.1 | - | 213 | 97 | |||
More Information
Set Membership: |
|

TTC4
Genomic Map