Assay Details
| Gene Symbol: RHOBTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144968.2 | XP_015000454.2 | 2095 | 94 | ||
| XM_015144970.2 | XP_015000456.2 | 2293 | 94 | |||
| XM_015144971.2 | XP_015000457.2 | 2078 | 94 | |||
| GenBank mRNA | JV636053.1 | - | 1846 | 94 | ||
| JV668211.1 | - | 2166 | 94 | |||
More Information
Set Membership: |
|

RHOBTB2
Genomic Map