Assay Details
| Gene Symbol: IARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117255.2 | XP_014972741.1 | 2281 | 68 | ||
| XM_015117256.2 | XP_014972742.1 | 2253 | 68 | |||
| GenBank mRNA | JU322638.1 | - | 2255 | 68 | ||
| JU475415.1 | - | 2234 | 68 | |||
| JV044249.1 | - | 2222 | 68 | |||
| JV505000.1 | - | 2155 | 68 | |||
| JV564976.1 | - | 2211 | 68 | |||
More Information
Set Membership: |
|

IARS1
Genomic Map