Assay Details
Gene Symbol: SYNGR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194830.3 | NP_001181759.1 | 579 | 69 | ||
XM_001113574.4 | XP_001113574.1 | 756 | 69 | |||
XM_015124377.2 | XP_014979863.1 | 797 | 69 | |||
XM_015124378.2 | XP_014979864.1 | 775 | 69 | |||
XM_028838487.1 | XP_028694320.1 | 524 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
