Assay Details
| Gene Symbol: ITGBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837273.1 | XP_028693106.1 | 1810 | 71 | ||
| GenBank mRNA | JV665845.1 | - | 1025 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ITGBL1| Gene Symbol: ITGBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837273.1 | XP_028693106.1 | 1810 | 71 | ||
| GenBank mRNA | JV665845.1 | - | 1025 | 71 | ||
Set Membership: |
|