Assay Details
| Gene Symbol: TGFBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112621.4 | XP_001112621.1 | 682 | 126 | ||
| XM_015117216.2 | XP_014972702.1 | 694 | 126 | |||
| XM_028835027.1 | XP_028690860.1 | 2783 | 126 | |||
| GenBank mRNA | JV722178.1 | - | 502 | 126 | ||
More Information
Set Membership: |
|

TGFBR1
Genomic Map