Assay Details
| Gene Symbol: RAB24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141506.2 | XP_014996992.1 | 619 | 60 | ||
| XM_015141507.2 | XP_014996993.1 | 554 | 60 | |||
| XM_015141508.2 | XP_014996994.1 | 525 | 60 | |||
| GenBank mRNA | JU319791.1 | - | 618 | 60 | ||
| JU472881.1 | - | 250 | 60 | |||
| JU476042.1 | - | 597 | 60 | |||
| JU476043.1 | - | 511 | 60 | |||
| JV047403.1 | - | 552 | 60 | |||
| JV047404.1 | - | 466 | 60 | |||
| JV490604.1 | - | 603 | 60 | |||
| JV528613.1 | - | 616 | 60 | |||
More Information
Set Membership: |
|

RAB24
Genomic Map