Assay Details
| Gene Symbol: INO80E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194364.2 | NP_001181293.1 | 232 | 64 | ||
| GenBank mRNA | JU471949.1 | - | 255 | 64 | ||
| JV712552.1 | - | 232 | 64 | |||
More Information
Set Membership: |
|

INO80E
Genomic Map