Assay Details
| Gene Symbol: EMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130274.2 | XP_014985760.1 | 881 | 70 | ||
| GenBank mRNA | JU319869.1 | - | 598 | 70 | ||
| JU472245.1 | - | 607 | 70 | |||
| JV047630.1 | - | 605 | 70 | |||
| JV529734.1 | - | 573 | 70 | |||
More Information
Set Membership: |
|

EMC3
Genomic Map