Assay Details
| Gene Symbol: FDPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111484.2 | XP_014966970.1 | 329 | 91 | ||
| XM_028828208.1 | XP_028684041.1 | 331 | 91 | |||
| GenBank mRNA | JU329863.1 | - | 60 | 91 | ||
| JU470993.1 | - | 190 | 91 | |||
More Information
Set Membership: |
|

FDPS
Genomic Map