Assay Details
| Gene Symbol: RMDN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261760.1 | NP_001248689.1 | 1065 | 63 | ||
| XM_015141885.2 | XP_014997371.2 | 1885 | 63 | |||
| XM_015141886.2 | XP_014997372.2 | 1880 | 63 | |||
| XM_015141887.2 | XP_014997373.1 | 1300 | 63 | |||
| XM_028850306.1 | XP_028706139.1 | 1169 | 63 | |||
| GenBank mRNA | JU345278.1 | - | 997 | 63 | ||
| JU476222.1 | - | 1065 | 63 | |||
More Information
Set Membership: |
|

RMDN3
Genomic Map