Assay Details
| Gene Symbol: TEAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136211.2 | XP_014991697.1 | 911 | 130 | ||
| XM_015136212.2 | XP_014991698.1 | 848 | 130 | |||
| XM_015136213.2 | XP_014991699.1 | 838 | 130 | |||
| XM_028847637.1 | XP_028703470.1 | 911 | 130 | |||
| XM_028847638.1 | XP_028703471.1 | 1993 | 130 | |||
| GenBank mRNA | JV659953.1 | - | 639 | 130 | ||
More Information
Set Membership: |
|

TEAD3
Genomic Map