Assay Details
| Gene Symbol: LOC695047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083464.4 | XP_001083464.3 | 140 | 97 | ||
| XM_015119019.2 | XP_014974505.2 | 1127 | 97 | |||
| XM_015119101.2 | XP_014974587.2 | 160 | 97 | |||
| XM_015119175.2 | XP_014974661.1 | 1127 | 97 | |||
| GenBank mRNA | JU471501.1 | - | 1012 | 97 | ||
| JU471503.1 | - | 1096 | 97 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015119101.2 transcripts.
Set Membership: |
|

LOC695047
Genomic Map