Assay Details
| Gene Symbol: INO80D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110775.2 | XP_014966261.1 | 2136 | 82 | ||
| XM_015110776.2 | XP_014966262.1 | 2164 | 82 | |||
| XM_015110777.2 | XP_014966263.1 | 2235 | 82 | |||
| XM_015110779.2 | XP_014966265.1 | 2144 | 82 | |||
| XM_015110780.2 | XP_014966266.1 | 2067 | 82 | |||
| XM_028831021.1 | XP_028686854.1 | 2124 | 82 | |||
More Information
Set Membership: |
|

INO80D
Genomic Map