Assay Details
| Gene Symbol: PPP1CC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257831.1 | NP_001244760.1 | 825 | 145 | ||
| XM_015152791.2 | XP_015008277.1 | 1077 | 145 | |||
| GenBank mRNA | JU335196.1 | - | 825 | 145 | ||
| JU335197.1 | - | 825 | 145 | |||
| JU470495.1 | - | 1975 | 145 | |||
| JU470496.1 | - | 1931 | 145 | |||
| JU470497.1 | - | 1979 | 145 | |||
| JV043673.1 | - | 900 | 145 | |||
| JV472877.1 | - | 722 | 145 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPP1CC
Genomic Map